Learn More
STEMCELL Technologies RoboSep Rat CD8+ T Cell Isolation Kit, For processing: 1 x 10 9 cells
SDP
Catalog No. 502390436
Encompass
The EasySep Rat CD8 T Cell Isolation Kit is designed to isolate CD8 T cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection. The EasySep procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep magnet and simply pouring or pipetting the desired cells into a new tube.
Catalog No. 50-239-0436 Supplier STEMCELL Technologies Supplier No. 19643RF
The EasySep Rat CD8 T Cell Isolation Kit is designed to isolate CD8 T cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection. The EasySep procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep magnet and simply pouring or pipetting the desired cells into a new tube.