Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Robotic Sterile Pipette Tips

Catalog No. 03411870 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
For Use With (Application):
For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 50 μL, Non-conductive, Material: clear polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
03-411-870 For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene
03-411-869 For: Tecan, Volume: 50 μL, Non-conductive, Material: clear polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-411-870 Supplier Bel-Art™ Supplier No. F378602100

Designed for use with automated liquid handling equipment

  • Sterile, disposable, filtered pipette tips can be used in robotic systems
    • Filtered to prevent contamination of pipette arm
  • Conductive tip models detect liquid level when pipetting conductive liquids
    • Ensures repeatable volume accuracy
  • DNase, RNase, pyrogen free, manufactured in Class 8 clean room
  • CE marked

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance CE marked
For Use With (Application) For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene
Format Hanging Tip Rack
Tip Style Standard
Packaging 16 racks x 96 tips
Quantity 1536
Product Type Robotic pipette tips
Conductivity No
Disposable Single-use
For Use With (Equipment) Tecan Genesis Freedom, Freedom EVO and Miniprep with LiHa
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.