Learn More
Fluotics Agilent Bravo compatible P10, sterile
Catalog No. NC1248631
Encompass_Preferred
Agilent Bravo compatible P10, sterile, working volume: 10 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 10734-312
Catalog No. NC1248631 Supplier Fluotics Supplier No. AGI10.ST
Agilent Bravo compatible P10, sterile, working volume: 10 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 10734-312