Learn More
AdipoGen Robotnikinin,Chemical. CAS: 1132653-79-2
SDP
Supplier:  AdipoGen AGCR10069M001
Encompass_Preferred
Robotnikinin, AdipoGen Life Sciences. Chemical. CAS: 1132653-79-2. Formula: C25H27ClN2O4. MW: 454.9. Synthetic. Sonic hedgehog (Shh) signaling modulator. Inhibits the Shh pathway in human cell lines upstream of Smo. Targets the Shh N-terminal protein.
Catalog No. 50-464-659