Learn More
Roboz Surgical Perforated Spoon, 5.25" (135 mm) long, 8 mm diameter, 1.5 mm depth, round handle.
SDP
Catalog No. NC1647594
Encompass
These shallow perforated spoons are used for handling organs or embryos. The perforations help drain fluids when the organs are removed from a solution.
Catalog No. NC1647594 Supplier Roboz Surgical Supplier No. RS6160
These shallow perforated spoons are used for handling organs or embryos. The perforations help drain fluids when the organs are removed from a solution.