Learn More
Roche Diagnostics KAPA2G Robust PCR Kits - HotStart PCR Kit, 2500 U
Catalog No. 501965278 Shop All Roche Diagnostics Products
Encompass
KAPA2G Robust PCR Kits contain a second-generation DNA polymerase engineered to solve inconsistent amplification across a broad range of amplicon types (GC- and AT-rich). The high performance of the DNA polymerase is ideally suited for challenging PCR applications and difficult samples, eliminating the need for optimization using multiple enzymes and protocols. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5278 Supplier Roche Diagnostics Supplier No. 07961120001
KAPA2G Robust PCR Kits contain a second-generation DNA polymerase engineered to solve inconsistent amplification across a broad range of amplicon types (GC- and AT-rich). The high performance of the DNA polymerase is ideally suited for challenging PCR applications and difficult samples, eliminating the need for optimization using multiple enzymes and protocols. *This item is non returnable.