Learn More
Ohiolumex Co Inc ROBUST OXYGEN PROBE, SS TIP 03
SDP
Supplier:  Ohiolumex Co Inc PSOXROB20
Encompass
Robust oxygen probe, Stainless Steel tip 03mm and 200mm in length, ca. 2m cable, with optical isolation.
Catalog No. NC1434164