Learn More
Medchemexpress LLC HY-N1343 1mg Medchemexpress, Robustine CAS:2255-50-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN13431MG
Encompass_Preferred
Medchemexpress, HY-N1343 1mg Robustine CAS:2255-50-7 Robustine, a furoquinoline alkaloid, from Dictamnus albus, exhibits inhibitory potency against human phosphodiesterase 5 (hPDE5A) in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2835