Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Robustine | 2255-50-7 | 215.20 | 5 MG
SDP

Catalog No. 5000414798
Encompass_Preferred

Robustine is a furoquinoline alkaloid isolated from Dictamnus albus. It demonstrates inhibitory potency against human phosphodiesterase 5 (hPDE5A) in vitro.

  • Furoquinoline alkaloid
  • Inhibits human phosphodiesterase 5 (hPDE5A)
  • Purity of 99.70%
  • Molecular formula C12H9NO3
  • Appears as a white to off-white solid
  • Target: PDE5A
  • Shipping at room temperature (continental US)
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-14798 Supplier Medchemexpress LLC Supplier No. HYN13435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Robustine is a furoquinoline alkaloid isolated from Dictamnus albus. It demonstrates inhibitory potency against human phosphodiesterase 5 (hPDE5A) in vitro.

  • Furoquinoline alkaloid
  • Inhibits human phosphodiesterase 5 (hPDE5A)
  • Purity of 99.70%
  • Molecular formula C12H9NO3
  • Appears as a white to off-white solid
  • Target: PDE5A
  • Shipping at room temperature (continental US)
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.