Learn More
Medchemexpress LLC Robustine | 2255-50-7 | 215.20 | C12H9NO3 | 10 MG
SDP
Catalog No. 5000295683
Encompass_Preferred
Robustine is a furoquinoline alkaloid (C12H9NO3) isolated from Dictamnus albus that exhibits inhibitory activity against human phosphodiesterase 5 (hPDE5A) in vitro. The compound is provided for research use with reported solubility in DMSO and recommended storage conditions for both powder and solutions.
  • Furoquinoline alkaloid with reported PDE5A inhibitory activity.
  • Small-molecule research compound (CAS 2255-50-7).
  • Soluble in DMSO (100 mg/mL) and formulatable in vitro at ≥2.5 mg/mL.
  • Stable when stored at 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes suitable for laboratory studies.
Catalog No. 50-002-95683 Supplier Medchemexpress LLC Supplier No. HYN134310MG
Robustine is a furoquinoline alkaloid (C12H9NO3) isolated from Dictamnus albus that exhibits inhibitory activity against human phosphodiesterase 5 (hPDE5A) in vitro. The compound is provided for research use with reported solubility in DMSO and recommended storage conditions for both powder and solutions.
  • Furoquinoline alkaloid with reported PDE5A inhibitory activity.
  • Small-molecule research compound (CAS 2255-50-7).
  • Soluble in DMSO (100 mg/mL) and formulatable in vitro at ≥2.5 mg/mL.
  • Stable when stored at 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes suitable for laboratory studies.