Learn More
Selleck Chemical LLC ROC-325
SDP
Catalog No. 5000001611
Encompass_Preferred
ROC-325 is an orally available novel inhibitor of lysosomal-mediated autophagy which diminishes AML cell viability with the IC50 range of 0 7-2 2 M
Catalog No. 50-000-01611 Supplier Selleck Chemical LLC Supplier No. S852725MG
ROC-325 is an orally available novel inhibitor of lysosomal-mediated autophagy which diminishes AML cell viability with the IC50 range of 0 7-2 2 M