Learn More
Medchemexpress LLC Rocaglamide | 84573-16-0 | 505.56 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19356100MG
Encompass_Preferred
Rocaglamide (Roc-A) is isolated from the genus *Aglaia* and is used for coughs, injuries, asthma, and inflammatory skin diseases. It acts as a potent inhibitor of NF-κB activation in T-cells. Rocaglamide is also a potent and selective heat shock factor 1 (HSF1) activation inhibitor with an IC50 of approximately 50 nM. Additionally, it inhibits the function of the translation initiation factor eIF4A and possesses anticancer properties in leukemia.
  • Isolated from the genus Aglaia.
  • Potent inhibitor of NF-κB activation in T-cells.
  • Potent and selective HSF1 activation inhibitor (IC50 of ~50 nM).
  • Inhibits the function of translation initiation factor eIF4A.
  • Exhibits anticancer properties in leukemia.
  • Can be used for coughs, injuries, asthma, and inflammatory skin diseases.
Catalog No. 50-003-85790