Learn More
Medchemexpress LLC Rocaglamide | 84573-16-0 | 505.56 | 50 MG
SDP
Catalog No. 5000385214
Encompass_Preferred
Rocaglamide is isolated from the genus Aglaia and can be used for coughs, injuries, asthma, and inflammatory skin diseases. It is a potent inhibitor of NF-κB activation in T-cells and a selective heat shock factor 1 (HSF1) activation inhibitor with an IC50 of ~50 nM. Rocaglamide also inhibits the function of the translation initiation factor eIF4A and exhibits anticancer properties in leukemia.
  • Isolated from the genus Aglaia.
  • Can be used for coughs, injuries, asthma, and inflammatory skin diseases.
  • Potent inhibitor of NF-κB activation in T-cells.
  • Potent and selective heat shock factor 1 (HSF1) activation inhibitor.
  • Inhibits the function of the translation initiation factor eIF4A.
  • Has anticancer properties in leukemia.
Catalog No. 50-003-85214 Supplier Medchemexpress LLC Supplier No. HY1935650MG
Rocaglamide is isolated from the genus Aglaia and can be used for coughs, injuries, asthma, and inflammatory skin diseases. It is a potent inhibitor of NF-κB activation in T-cells and a selective heat shock factor 1 (HSF1) activation inhibitor with an IC50 of ~50 nM. Rocaglamide also inhibits the function of the translation initiation factor eIF4A and exhibits anticancer properties in leukemia.
  • Isolated from the genus Aglaia.
  • Can be used for coughs, injuries, asthma, and inflammatory skin diseases.
  • Potent inhibitor of NF-κB activation in T-cells.
  • Potent and selective heat shock factor 1 (HSF1) activation inhibitor.
  • Inhibits the function of the translation initiation factor eIF4A.
  • Has anticancer properties in leukemia.