Learn More
MilliporeSigma ROCare A
Catalog No. ZWACIDC12 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 Packages
Catalog No. Quantity
ZWACIDC12 12 Packages
1 options
Catalog No. ZWACIDC12 Supplier MilliporeSigma Supplier No. ZWACIDC12
Add to Cart
Add to Cart
Acid cleaning chemical removes scaling from RO membranes
  • ROCare A is an acidic cleaner that protects the water purification systems by effectively removing some or most mineral scale buildup on the surface of the reverse osmosis (RO) membranes.
  • It is formulated with citric acid only, significantly reducing toxicity, ensuring safety and supporting environmental sustainability.
  • The acid cleaner is delivered in a pouch and the empty pouch is removed after use.
  • It is to be used periodically when prompted by the system, when the RO % rejection drops more than 5%, or when the RO permeate flow rate drops more than 10% due to mineral scaling.
  • The ROCare A pouch cleans RO membranes supporting Milli-Q™ HR/HX/CLX, Elix™ Large, Milli-Q™ IX, Milli-Q™ IQ, Milli-Q™ EQ, Elix™ Advantage, Milli-Q™ Integral and Essential water purification systems.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 12 Packages
Type Anti-scaling Agent