Learn More
Med Vet International Miltex Vantage Rochester-Ochsner Forceps, 7.25in, Straight
Supplier:  Med Vet International MILV97152
Encompass_Preferred
The Miltex Vantage V97-152 Rochester Ochsner Forcepshave a length of 7.25in. and are made of Mid-Grade stainlesss steel. They have a straight tip and are commonly used to clamp off blood vessels to avoid bleeding during surgical procedures. These forceps have 1x2 teeth.
Catalog No. 50-255-9432