Learn More
Med Vet International Miltex Vantage Rochester-Pean Forceps, 9in., Straight
Catalog No. 502831607
Encompass_Preferred
The Miltex Vantage V97-126 Rochester-Pean Forceps have a length of 9in. and are made of mid-grade stainless steel. They have a straight tip and are a hemostatic forcep with fully serrated jaws. This surgical forcep is commonly used to occlude or clamp blood vessels to control bleeding during surgery.
Catalog No. 50-283-1607 Supplier Med Vet International Supplier No. MILV97126
The Miltex Vantage V97-126 Rochester-Pean Forceps have a length of 9in. and are made of mid-grade stainless steel. They have a straight tip and are a hemostatic forcep with fully serrated jaws. This surgical forcep is commonly used to occlude or clamp blood vessels to control bleeding during surgery.