Learn More
Roche Diagnostics ACCU-CHEK Inform II Controls
Catalog No. 22045963 Shop All Roche Diagnostics Products
Change view
Click to view available options
Content And Storage:
4°C to 30°C
Catalog No. Content And Storage
22-045-963 4°C to 30°C
1 options
This item is not returnable. View return policy
Catalog No. 22-045-963 Supplier Roche Diagnostics Supplier No. 05213509001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Can be used with ACCU-CHEK™ Inform II system with ACCU-CHEK Inform II test strips for performance
Control pack contains two control solutions
  • One for hypoglycemic range (control solution 1, gray cap)
  • One for hyperglycemic range (control solution 2, white cap)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C to 30°C
Type Control Pack