Learn More
Roche Diagnostics ACCU-CHEK Inform II Linearity Test Kit
Catalog No. 22045964 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
6 x 2.5 mL
Catalog No. Quantity
22-045-964 6 x 2.5 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-045-964 Supplier Roche Diagnostics Supplier No. 05871166001
Add to Cart
Add to Cart
This item is not returnable. View return policy
For periodic verification of linearity of ACCU-CHEK Inform II system using ACCU-CHEK Inform II test strips
  • Contains six linearity solutions with varying glucose levels, linearity logs and inserts
  • 6 x 2.5mL linearity solutions
  • For professional use only
TRUSTED_SUSTAINABILITY
Specifications
Type Linearity Test Kit
Quantity 6 x 2.5 mL