Learn More
Roche Diagnostics ACCU-CHEK™ Safe-T-Pro™ Lancets
Catalog No. 1367845 Shop All Roche Diagnostics Products
Change view
Click to view available options
Safe-T-Pro Lancets
Catalog No.
13-678-45 Safe-T-Pro Lancets
1 options
This item is not returnable. View return policy
Catalog No. 13-678-45 Supplier Roche Diagnostics Supplier No. 03136752001
This item is not returnable. View return policy
Designed with Safe-T-Wings tabs for patient and user protection from reuse or accidental contact
  • Single-use, disposable lancet for taking fingerstick samples of whole blood
  • T-shape makes handling easier
  • When blue trigger button is pressed to obtain a sample, spring-loaded firing action breaks the lancet wings to provide a superior fail-safe against reuse
  • For added protection, lancet permanently retracts into its protective case after use to prevent accidental needlesticks and contamination
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
For Use With (Application) For patient and user protection from reuse or accidental contact
Quantity 200 Pack
Style Safe-T-Pro™ lancets
Includes Spring-loaded, Safe-T-Wings tabs
Product Type ACCU-CHEK™ Safe-T-Pro™ Lancet