Learn More
Roche Diagnostics ACCU-CHEK Safe-T-Pro Plus Lancet
Catalog No. 22045102 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
200/Pk.
Catalog No. Quantity
22-045-102 200/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-045-102 Supplier Roche Diagnostics Supplier No. 03448622001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type ACCU-CHEK™ Safe-T-Pro™ Lancet
Model Safe-T-Pro Plus
Includes Three penetration depth settings, permanent needle retraction
Quantity 200/Pk.
Style Safe-T-Pro™ Plus lancets