Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics AVL 9180 Series Electrolyte Analyzers

Catalog No. avlgd5043 Shop All Roche Diagnostics Products
Change view
Click to view available options
Sample Type:
Whole Blood, Plasma, Serum, Acetate or Bicarbonate Dialysate or Pre-diluted Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
AVLGD5043 Whole Blood, Plasma, Serum, Acetate or Bicarbonate Dialysate or Pre-diluted Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AVLGD5043 Supplier Roche Diagnostics Supplier No. 03157334001
Only null left
Add to Cart
Add to Cart
Request a Quote

Includes

Complete with reference electrode, reference housing, sodium electrode, accessory kit, initial supply of calibrating and operating solutions, ISE-trol Electrolyte Control, printer paper, extra maintenance supplies, and operator's video and manual

Stand-alone, fully automatic analyzer with nine interchangeable electrolyte configurations

  • 95μL whole blood, serum, plasma, acetate or bicarbonate dialysate, or pre-diluted urine
  • Simple configuration with Yes/No dialog—no need to change reagents or replace tubing
  • Accepts collection tubes, syringes, capillaries or cuvettes
  • All reagents and waste are sealed in convenient SnapPak™
  • Monitors remaining reagent and prompts user to insert a fresh SnapPak when required
  • One-point calibration with each test; two-point, every four hours
  • On-board QC software tracks QC data, including standard deviation and C.V.
  • Stand-by mode during idle periods
  • Built-in thermal printer

Certifications

CE marked CSA/CE approved

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Whole Blood, Plasma, Serum, Acetate or Bicarbonate Dialysate or Pre-diluted Urine
For Use With (Application) Measures electrolytes from whole Blood, serum, plasma, urine dialysate or aqueous standards
Analysis Time 50 sec.
Sample Size 95 μL
Humidity 5 to 85% Non-condensing
Calibration Yes
Certifications/Compliance CE, CSA/CE
Depth (English) 12 in.
Depth (Metric) 304.8 mm
Height (English) 13.2 in.
Height (Metric) 335 mm
Width (English) 12.4 in.
Width (Metric) 315 mm
Electrical Requirements 100/240 V 50/60 Hz
Amperage 1.4 A
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.