Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Chemstrip Micral Albumin Test

Catalog No. 23305022 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
30 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-305022 30 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-305022 Supplier Roche Diagnostics Supplier No. 11544039160
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Two easy steps — results in one minute.

  • A semi-quantitative visual immunoassay test that is specific for human albumin in urine
  • The only “dipstick” test for MAU that can be used in any office
  • Convenient for both patient and physician
  • Five-minute color stability
  • Specificity and sensitivity: 94%
  • CLIA waived

TRUSTED_SUSTAINABILITY

Specifications

Clia Complexity Waived
Type Microalbumin Reagent Strip
For Use With (Application) Semi-quantitative visual immunoassay test that is specific for Human Albumin in Urine
Quantity 30 Pack
Test Time 1 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.