Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Cholesterol Controls for Accutrend Plus Meters

Catalog No. 22045732 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
2 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-045-732 2 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-045-732 Supplier Roche Diagnostics Supplier No. 05219957001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Cholesterol Control–Low Level
  • Cholesterol Control–High Level
  • 2mL each

Cholesterol control solutions allow you to perform quality control checks to ensure that your system is working properly

Each control solution contains a known amount of cholesterol

  • Package provides one bottle of each low level and high level cholesterol control solutions.
  • The soultions are used exactly in the same way as a drop of blood.
  • If the control soultion is within an acceptable range, you know that your meter and test srtips are working properly and that you are performing the tests correctly.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 25°C
For Use With (Equipment) Accutrend Plus™ system
Type Accutrend Cholesterol Control
Includes Cholesterol control low level, Cholesterol control high level, 2mL each
Quantity 2 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.