Learn More
Roche Diagnostics Capillary Tubes for use with CoaguChek XS System
Catalog No. 23312188 Shop All Roche Diagnostics Products
Change view
Click to view available options
For Use With (Equipment):
CoaguChek XS System
Catalog No. For Use With (Equipment)
23-312188 CoaguChek XS System
1 options
This item is not returnable. View return policy
Catalog No. 23-312188 Supplier Roche Diagnostics Supplier No. 11621173001
Add to Cart
Add to Cart
This item is not returnable. View return policy
100 Plastic Disposable Capillary Tubes, 10 Bulbs
  • Capillary tube designed to easily collect blood and precisely apply to test strip
  • Suited for use with Roche's CoaguChek XS blood collection system
  • Disposable for savings and infection prevention
The CoaguChek XS System (04837975001) is a portable, extremely efficient instrument used by doctors and healthcare professionals to monitor patients under anticoagulant therapy. The System is less invasive, improves patient compliance and offers the advantage of immediate therapy adjustment.
Order Info
Roche provides technical assistance 24 hours-a-day.
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Tubes and Bulbs
For Use With (Equipment) CoaguChek XS System
Includes Pack of 100 Plastic Disposable Capillary Tubes, 10 Bulbs.