Description
Includes
CoaguChek XS Plus meter, test strip guide cover, power adapter, universal battery pack, battery compartment cover, starshaped screwdriver, user manual and getting started guide.
Requires
PT strips (in a vial): Box with 2 vials of 24 strips each with a code chip; Box with 1 vial of 24 strips with a code chip.
The professional solution for clinical practices.
Accurate and flexible technology — the smart way to test
- Accurate, precise results in about one minute
- The only system to perform onboard quality control and determine patient results in a single test chamber
- Neutralizes therapeutic levels of heparin and LMWH
- INR corrected for hematocrit within specified range
- Built-in barcode scanner for operator IDs and patient IDs
- Stores 1000 patient and 500 QC test results
- Operator and QC lockout capabilities
- Code chip programming
- Data storage for up to 60 code chips
Easy and convenient to use
- Small sample size (8μL) from a fingerstick
- Top or side strip dosing, with outside-meter blood application (avoiding potential cross-contamination)
- New onboard controls — no external QC necessary
- On-the-spot adjustment of your patient's VKA dosage — no need to wait for lab results
Specifications
Specifications
| Type | XS Plus System |
| Includes | CoaguChek XS Plus instrument, handheld power supply unit, printed manual in English, CD-ROM with other languages: German, French, Italian, English, Spanish, Portuguese, Danish, Norwegian, Swedish, Finnish and Dutch |