Learn More
Roche Diagnostics CoaguChek™ XS Plus System
Catalog No. 23312197 Shop All Roche Diagnostics Products
Change view
Click to view available options
Type:
CoaguChek Handheld Base Unit
XS Plus System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-312-197 CoaguChek Handheld Base Unit
22-044-750 XS Plus System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-312-197 Supplier Roche Diagnostics Supplier No. 08832366001
Add to Cart
Add to Cart
Built-in QC and data management for POC anticoagulation testing
The professional solution for clinical practices.
Accurate and flexible technology — the smart way to test
  • Accurate, precise results in about one minute
  • The only system to perform onboard quality control and determine patient results in a single test chamber
  • Neutralizes therapeutic levels of heparin and LMWH
  • INR corrected for hematocrit within specified range
Streamlined data management
  • Stores 1000 patient and 500 QC test results
  • Operator and QC lockout capabilities
  • Code chip programming
  • Data storage for up to 60 code chips
Easy and convenient to use
  • Small sample size (8μL) from a fingerstick
  • Top or side strip dosing, with outside-meter blood application (avoiding potential cross-contamination)
  • New onboard controls — no external QC necessary
  • On-the-spot adjustment of your patient's VKA dosage — no need to wait for lab results
TRUSTED_SUSTAINABILITY
Specifications
Type CoaguChek Handheld Base Unit
For Use With (Equipment) CoaguChek XS Plus Meter, CoaguChek XS Pro meter
Includes CoaguChek XS Plus instrument, handheld power supply unit, printed manual in English, CD-ROM with other languages: German, French, Italian, English, Spanish, Portuguese, Danish, Norwegian, Swedish, Finnish and Dutch
Quantity 1
Height (English) 3.22 in.
Height (Metric) 8.18 cm
Width (English) 4.72 in.
Width (Metric) 11.99 cm