Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-CHEK Coagulation Lancets

Catalog No. 22045105 Shop All Roche Diagnostics Products
Change view
Click to view available options
Lancet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-045-105 Lancet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-045-105 Supplier Roche Diagnostics Supplier No. 08029016160

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coagulation Lancet Needle, Multiple Depth Settings, 23 Gauge

  • Color: purple
  • Multiple depth settings
  • Retractable
  • Incision type: needle
  • 23 gauge
  • Sterile
  • Target area: Fingertip
  • Disposable
TRUSTED_SUSTAINABILITY

Specifications

Depth (English) Multiple Depth Settings
For Use With (Application) Coagulation Testing
Quantity 200/Pk.
Product Type Lancet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.