Learn More
Roche Diagnostics Urisys™ 1100 Urinalysis Analyzer
Catalog No. 22044792 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
22-044-792 Each
1 options
Catalog No. 22-044-792 Supplier Roche Diagnostics Supplier No. 03617556001
Add to Cart
Add to Cart
Semi-quantitative analyzer for evaluation of Chemstrip™ urine test strips
  • The CLIA-waived Urisys 1100 urine analyzer performs an automated read-out of Roche's ten-parameter Chemstrip 10 MD with results in as fast as 35 seconds and a throughput of 45 urine samples per hour.
  • Trusted Performance: Additional safety features include QC management (with automated QC lock-out when necessary), user management, strip lot management and calibration management.
  • Improved Usability: The redesigned Urisys 1100 launched in 2024 features a 4.3 in. color touchscreen, an internal memory of 1,000 patient results and the ability to update software remotely via USB WiFi stick (sold separately; not a Roche product) or LAN connection.
  • Ready to Connect: Transfer patient results to your preferred data management system via USB WiFi stick or LAN connection.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Analyzer
For Use With (Equipment) Chemstrip™ Urine Test Strip
Quantity Each