Learn More
Description
- Manufactured from medical grade polypropylene
- Half-skirt feature
Specifications
Specifications
| Barcode | No Barcode |
| Array | 8 x 12 |
| Color | Natural |
| Product Type | PCR Microplate |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| For Use With (Application) | Roche LightCycler PCR |
| Quantity | 100/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.