Learn More
Hach Company Rochelle Salt Solution, 500 mL
Supplier:  Hach Company 172549
Encompass
Potassium sodium tartrate solution. Used to mask hardness interference in some analytical methods.
Catalog No. 50-199-4590