Learn More
Med Vet International Miltex Vantage Rochester-Carmalt Forceps, 8in., Straight
Catalog No. 502831174
Encompass_Preferred
The Miltex Vantage V97-180 Rochester-Carmalt Forceps have a length of 8in. and are made of mid-grade stainless steel. They have a straight tip and are a great hemostat for clamping blood vessels and grasping large tissues. These forceps have a textured blade and scissor like ratchet handle that ensures a strong grip with minimal chance of cutting the tissue.
Catalog No. 50-283-1174 Supplier Med Vet International Supplier No. MILV97180
The Miltex Vantage V97-180 Rochester-Carmalt Forceps have a length of 8in. and are made of mid-grade stainless steel. They have a straight tip and are a great hemostat for clamping blood vessels and grasping large tissues. These forceps have a textured blade and scissor like ratchet handle that ensures a strong grip with minimal chance of cutting the tissue.