Learn More
Sklar Instruments Forceps, Merit Rochester-Pean Hemostatic Forceps, Sklar 8 in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126630 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Rochester-Pean Hemostatic Forceps, Sklar, 8 in., Straight, Serrated, Stainless Steel, EA. Merit Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with full, horizontally serrated jaws, and a length of 8 inches.
Catalog No. 50-112-6630 Supplier Sklar Instruments Supplier No. 97492
Forceps, Merit Rochester-Pean Hemostatic Forceps, Sklar, 8 in., Straight, Serrated, Stainless Steel, EA. Merit Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with full, horizontally serrated jaws, and a length of 8 inches.