Learn More
Sklar Instruments Rochester-Ochsner (Kocher) Forceps, Sklar, 6-1/4 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel.
SDP
Catalog No. 501437405 Shop All Sklar Instruments Products
Encompass_Preferred
Rochester-Ochsner (Kocher) Forceps, Sklar, 6-1/4 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar Rochester-Oschner Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They have horizontal serrations the entire length of the jaw as well as 1 X 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. This product is straight with full, horizontally serrated jaws, 1 X 2 teeth, and a length of 6-1/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7405 Supplier Sklar Instruments Supplier No. 172862
Rochester-Ochsner (Kocher) Forceps, Sklar, 6-1/4 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar Rochester-Oschner Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They have horizontal serrations the entire length of the jaw as well as 1 X 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. This product is straight with full, horizontally serrated jaws, 1 X 2 teeth, and a length of 6-1/4 inches.