Learn More
Enterprise Technology Solutions Rochester Midland Corporation Cleaner,f/Toilet&Bathroom,Descaler,Tamer,Foam,1QT,6/CT,Blue Green
SDP
Catalog No. 5000830157
Encompass_Preferred
Thickened, tamed-acid cleaner effectively cleans, descales, derusts and polishes toilet bowls, showers and more. Foaming cleaning solution matches powerful, traditional hydrochloric acid cleaners. However, unique added ingredients 'tame' potential harm for both product users and those who come in contact with affected surfaces for safer cleaning. Use to clean shower rooms, restrooms and other large areas. Cleaner leaves behind refreshing, floral scent.
Catalog No. 50-008-30157 Supplier Enterprise Technology Solutions Supplier No. RCM11811214
Thickened, tamed-acid cleaner effectively cleans, descales, derusts and polishes toilet bowls, showers and more. Foaming cleaning solution matches powerful, traditional hydrochloric acid cleaners. However, unique added ingredients 'tame' potential harm for both product users and those who come in contact with affected surfaces for safer cleaning. Use to clean shower rooms, restrooms and other large areas. Cleaner leaves behind refreshing, floral scent.