Learn More
Enterprise Technology Solutions Rochester Midland Corporation Cleaner Refill,All-Purpose,Low-Foam,EZ-Mix,64.25oz,4/CT,GN
SDP
Catalog No. 5000789812
Encompass_Preferred
Enviro Care Low-foam All-purpose Cleaner is highly concentrated and great for general-purpose cleaning. Bio-based, neutral pH is ideal for removing soils from floors and hard surfaces and floor cleaning through automatic scrubbers. Cleaner automatically dilutes through RMC's EZ Mix Dispenser System (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.
Catalog No. 50-007-89812 Supplier Enterprise Technology Solutions Supplier No. RCM11822099
Enviro Care Low-foam All-purpose Cleaner is highly concentrated and great for general-purpose cleaning. Bio-based, neutral pH is ideal for removing soils from floors and hard surfaces and floor cleaning through automatic scrubbers. Cleaner automatically dilutes through RMC's EZ Mix Dispenser System (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.