Learn More
Enterprise Technology Solutions Rochester Midland Corporation DFE Sabre, Bio-Catalytic, 1.9L, 4/CT, White
SDP
Catalog No. 5000789819
Encompass_Preferred
Bio-catalytic cleaner/degreaser breaks down fats, oils and greases. It provides long-term enhanced cleaning to keep floors and grout spotless while reducing risk of slip and falls. DfE Sabre is great for foodservice, kitchen and restroom floors. Cleaner automatically dilutes through RMC's EZ Mix Dispenser (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.
Catalog No. 50-007-89819 Supplier Enterprise Technology Solutions Supplier No. RCM11974099
Bio-catalytic cleaner/degreaser breaks down fats, oils and greases. It provides long-term enhanced cleaning to keep floors and grout spotless while reducing risk of slip and falls. DfE Sabre is great for foodservice, kitchen and restroom floors. Cleaner automatically dilutes through RMC's EZ Mix Dispenser (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.