Learn More
Enterprise Technology Solutions Rochester Midland Corporation Enviro Care Washroom Cleaner E-Z Mix, 1.9L, 4/CT, GN
SDP
Catalog No. 5000829707
Encompass_Preferred
Bio-based washroom cleaner offers a mild acid formulation that removes soap film, body oils and mineral deposits from any washable surface, such as toilets, urinals and bathroom fixtures. Nontoxic, phosphate-free cleaner removes difficult stains from ceramic tile floors and grouting. Cleaner automatically dilutes through RMC's EZ Mix Dispenser (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.
Catalog No. 50-008-29707 Supplier Enterprise Technology Solutions Supplier No. RCM12002099
Bio-based washroom cleaner offers a mild acid formulation that removes soap film, body oils and mineral deposits from any washable surface, such as toilets, urinals and bathroom fixtures. Nontoxic, phosphate-free cleaner removes difficult stains from ceramic tile floors and grouting. Cleaner automatically dilutes through RMC's EZ Mix Dispenser (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.