Learn More
Enterprise Technology Solutions Rochester Midland Corporation Glass Cleaner, Bio Based, Measurement Sys, 32 oz, Purple
SDP
Catalog No. 5000789848
Encompass_Preferred
Bio-based glass cleaner is a convenient, accurate and green complete solution for your glass cleaning needs. Enviro mix packaging allows you to accurately and easily mix concentrates with a built-in measurement system in a designated spray bottle (sold separately). Simply squeeze and fill the upper chamber with concentrate to then pour the cleaner into your spray bottle and fill with water for easy cleaning. Enviro Care Glass Cleaner leaves glass and mirrors streak-free. Use also on enamel, stainless steel, chrome, formica, plastic tile, ceramic tile and other polished surfaces. VOC-compliant formula contains no ammonia or alcohol. Dilution ratio is 1 oz. of cleaner per 31 oz. of water.
Catalog No. 50-007-89848 Supplier Enterprise Technology Solutions Supplier No. RCM12001014
Bio-based glass cleaner is a convenient, accurate and green complete solution for your glass cleaning needs. Enviro mix packaging allows you to accurately and easily mix concentrates with a built-in measurement system in a designated spray bottle (sold separately). Simply squeeze and fill the upper chamber with concentrate to then pour the cleaner into your spray bottle and fill with water for easy cleaning. Enviro Care Glass Cleaner leaves glass and mirrors streak-free. Use also on enamel, stainless steel, chrome, formica, plastic tile, ceramic tile and other polished surfaces. VOC-compliant formula contains no ammonia or alcohol. Dilution ratio is 1 oz. of cleaner per 31 oz. of water.