Learn More
Enterprise Technology Solutions Rochester Midland Corporation Glass Cleaner Spray Bottle, 1 Qt, Clear Frosted
SDP
Catalog No. 5000789825
Encompass_Preferred
Trigger-spray bottle is designed for use with RMC Enviro Care Glass Cleaner. Empty, silk-screened bottle already has a glass cleaner label around the base and specifies that contents are ready to use. Bottle is made of plastic. Design also includes a ringed neck for easy gripping. Trigger-sprayer top (sold separately) unscrews easily for refilling.
Catalog No. 50-007-89825 Supplier Enterprise Technology Solutions Supplier No. RCM35064373
Trigger-spray bottle is designed for use with RMC Enviro Care Glass Cleaner. Empty, silk-screened bottle already has a glass cleaner label around the base and specifies that contents are ready to use. Bottle is made of plastic. Design also includes a ringed neck for easy gripping. Trigger-sprayer top (sold separately) unscrews easily for refilling.