Learn More
Enterprise Technology Solutions Rochester Midland Corporation Neutral Cleaner/Degreaser,1.9L,Lavender,4/CT,PE
SDP
Catalog No. 5000830015
Encompass_Preferred
Perfecto 7 delivers exceptional grease-cutting properties that rapidly penetrate and emulsify a wide variety of soils to leave surfaces sparkling clean. The low-foam, neutral pH, all-purpose cleaner is formulated to clean walls, floors, chrome, porcelain and stainless steel. Neutral cleaner/degreaser has a long-lasting lavender fragrance. Cleaner automatically dilutes through RMC's EZ Mix Dispenser (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.
Catalog No. 50-008-30015 Supplier Enterprise Technology Solutions Supplier No. RCM11974199
Perfecto 7 delivers exceptional grease-cutting properties that rapidly penetrate and emulsify a wide variety of soils to leave surfaces sparkling clean. The low-foam, neutral pH, all-purpose cleaner is formulated to clean walls, floors, chrome, porcelain and stainless steel. Neutral cleaner/degreaser has a long-lasting lavender fragrance. Cleaner automatically dilutes through RMC's EZ Mix Dispenser (sold separately) that dispenses the right amount of product for spray bottles or buckets/auto scrubbers. EcoLogo is certified for reduced environmental impact.