Learn More
Sklar Instruments Forceps, Surgi-OR Rochester-Oschner (Kocher) Forceps, Sklar 7-1/4 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel, EA.
SDP
Catalog No. 501126542 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Surgi-OR Rochester-Oschner (Kocher) Forceps, Sklar, 7-1/4 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel, EA.Surgi-OR Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They have horizontal serrations the entire length of the jaw as well as 1 X 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6542 Supplier Sklar Instruments Supplier No. 95521
Forceps, Surgi-OR Rochester-Oschner (Kocher) Forceps, Sklar, 7-1/4 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel, EA.Surgi-OR Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They have horizontal serrations the entire length of the jaw as well as 1 X 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Lifetime warranty against all manufacturer defects.