Learn More
Sklar Instruments Rochester-Pean Forceps, Sklar, 10-1/4 in., Curved, Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501438269 Shop All Sklar Instruments Products
Encompass_Preferred
Rochester-Pean Forceps, Sklar, 10-1/4 in., Curved, Serrated, Stainless Steel. Sklar Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. This product is curved with full, horizontally serrated jaws, and a length of 10-1/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-8269 Supplier Sklar Instruments Supplier No. 362810
Rochester-Pean Forceps, Sklar, 10-1/4 in., Curved, Serrated, Stainless Steel. Sklar Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. This product is curved with full, horizontally serrated jaws, and a length of 10-1/4 inches.