Learn More
Electron Microscopy Sciences EMS Rochester Pean Forceps with Straight, 10-1/4"
SDP
Catalog No. 501931485
Encompass_Preferred
These forceps feature a finger ring grip and adjustable interlocking cross arms for tight clamping of tissue. They have serrated tips made of stainless steel.
Tips: Straight
OAL: 10-1/4"
Catalog No. 50-193-1485 Supplier Electron Microscopy Sciences Supplier No. 7299906
These forceps feature a finger ring grip and adjustable interlocking cross arms for tight clamping of tissue. They have serrated tips made of stainless steel.
Tips: Straight
OAL: 10-1/4"