Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rociletinib hydrobromide | 1446700-26-0 | MFCD28386290 | 98.0% | 636.46 g/mol | C27H29BrF3N7O3 | 10 MG
SDP

Catalog No. 5000287869
Encompass_Preferred

Rociletinib hydrobromide is the hydrobromide salt of rociletinib, an orally active kinase inhibitor selective for mutant forms of the epidermal growth factor receptor (EGFR) including T790M. It is supplied as a solid research reagent for biochemical, cellular, and analytical studies.

  • Hydrobromide salt of rociletinib.
  • Selective inhibitor of mutant EGFR, including T790M.
  • Intended for research and analytical applications.
  • Purity 98.04% (HPLC).
  • Molecular weight 636.46 g/mol.
  • CAS number 1446700-26-0.
  • Store sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-87869 Supplier Medchemexpress LLC Supplier No. HY15729A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rociletinib hydrobromide is the hydrobromide salt of rociletinib, an orally active kinase inhibitor selective for mutant forms of the epidermal growth factor receptor (EGFR) including T790M. It is supplied as a solid research reagent for biochemical, cellular, and analytical studies.

  • Hydrobromide salt of rociletinib.
  • Selective inhibitor of mutant EGFR, including T790M.
  • Intended for research and analytical applications.
  • Purity 98.04% (HPLC).
  • Molecular weight 636.46 g/mol.
  • CAS number 1446700-26-0.
  • Store sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.