Learn More
Medchemexpress LLC HY-15729A 5mg Medchemexpress, Rociletinib hydrobromide CAS:1446700-26-0 Purity:>98%
SDP
Catalog No. 502081780
Encompass_Preferred
Medchemexpress, HY-15729A 5mg Rociletinib hydrobromide CAS:1446700-26-0 Rociletinib hydrobromide (CO-1686 hydrobromide) is an orally delivered kinase inhibitor that specifically targets the mutant forms of EGFR including T790M, and the Ki values for EGFRL858R/T790M and EGFRWT are 21.5 nM and 303.3 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1780 Supplier Medchemexpress LLC Supplier No. HY15729A5MG
Medchemexpress, HY-15729A 5mg Rociletinib hydrobromide CAS:1446700-26-0 Rociletinib hydrobromide (CO-1686 hydrobromide) is an orally delivered kinase inhibitor that specifically targets the mutant forms of EGFR including T790M, and the Ki values for EGFRL858R/T790M and EGFRWT are 21.5 nM and 303.3 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.