Learn More
Apexbio Technology LLC Rocilinostat (ACY-1215) 1316214-52-4 200mg
SDP
Catalog No. 5000064324
Encompass_Preferred
Rocilinostat (ACY-1215 CAS 1316214-52-4) is a selective inhibitor of histone deacetylase 6 (HDAC6) HDAC6 enzymatically regulates various cellular processes including protein trafficking cell-cycle progression and cytoskeletal organization and its dysregulated activity is implicated in tumor progression and cancer metastasis Rocilinostat potently inhibits HDAC6 (IC50 5 nM) while showing minimal activity against other HDAC isoforms and sirtuins (IC50 1 M) Preclinical studies using multiple myeloma (MM) cell lines and xenograft mouse models demonstrated rocilinostat can enhance acetylation of -tubulin disrupt protein stability pathways and sensitize cancer cells to proteasome inhibitors such as bortezomib and carfilzomib resulting in increased apoptosis and reduction in tumor growth
Catalog No. 50-000-64324 Supplier Apexbio Technology LLC Supplier No. A4083200
Rocilinostat (ACY-1215 CAS 1316214-52-4) is a selective inhibitor of histone deacetylase 6 (HDAC6) HDAC6 enzymatically regulates various cellular processes including protein trafficking cell-cycle progression and cytoskeletal organization and its dysregulated activity is implicated in tumor progression and cancer metastasis Rocilinostat potently inhibits HDAC6 (IC50 5 nM) while showing minimal activity against other HDAC isoforms and sirtuins (IC50 1 M) Preclinical studies using multiple myeloma (MM) cell lines and xenograft mouse models demonstrated rocilinostat can enhance acetylation of -tubulin disrupt protein stability pathways and sensitize cancer cells to proteasome inhibitors such as bortezomib and carfilzomib resulting in increased apoptosis and reduction in tumor growth