Learn More
Apexbio Technology LLC Rocilinostat (ACY-1215), 50mg. Cas: 1316214-52-4 MFCD: MFCD22666356. Selective HDAC6 inhibitor
SDP
Supplier:  Apexbio Technology LLC A408350
Encompass_Preferred
Rocilinostat (ACY-1215) is a selective inhibitor of HDAC6 with IC50 of 5 nM [1].HDAC6 (histone deacetylase 6) is an enzyme which encoded by the HDAC6 gene and plays an important role in translational regulation, cell cycle progression and developmental events. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4874
 Restricted Item