Learn More
Cell Biolabs Inc 96-Well ROCK (Rho Kinase) Activity Assay Kit, 96 assays
Catalog No. NC9001833
Encompass_Preferred
Cell Biolabs’ 96-well ROCK Activity Assay Kit is an enzyme immunoassay developed for detection of the specific phosphorylation of MYPT1 at Thr696 by Rho-Associated Kinase (ROCK). A strip well microtiter plate is precoated with a recombinant MYPT1. After incubating the substrate wells with ROCK samples (such as purified kinase, cell or tissue lysate) the phosphorylated MYPT1 is detected by an anti-phospho-MYPT1 (Thr696) antibody. The kit has detection sensitivity limit of 200 pg of active ROCK-II. A recombinant active ROCK-II is also provided as a positive control. Each kit provides sufficient quantities to perform up to 96 assays.
Catalog No. NC9001833 Supplier Cell Biolabs Inc Supplier No. STA416
Cell Biolabs’ 96-well ROCK Activity Assay Kit is an enzyme immunoassay developed for detection of the specific phosphorylation of MYPT1 at Thr696 by Rho-Associated Kinase (ROCK). A strip well microtiter plate is precoated with a recombinant MYPT1. After incubating the substrate wells with ROCK samples (such as purified kinase, cell or tissue lysate) the phosphorylated MYPT1 is detected by an anti-phospho-MYPT1 (Thr696) antibody. The kit has detection sensitivity limit of 200 pg of active ROCK-II. A recombinant active ROCK-II is also provided as a positive control. Each kit provides sufficient quantities to perform up to 96 assays.