Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rock Formation and Identification Lab Investigation
SCI_ED

Catalog No. s32681
Change view
Click to view available options
Rock Formation and Identfctn Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S32681 Rock Formation and Identfctn Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S32681 Supplier Neo/Sci Corporation Supplier No. 201523

Includes

Set of rock specimens

Observe the distinct characteristics of each type of rock.

  • Explore the igneous, sedimentary, and metamorphic processes of rock formation

Order Info

Class size: 40 students.

Specifications

For Use With (Application) Observe the distinct characteristics of each type of rock
Product Type Rock Formation and Identification Kit
Sufficient For 40 Students
Includes Set of rock specimens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.