Learn More
Medchemexpress LLC HY-10319 5mg Medchemexpress, ROCK-IN-2 CAS:867017-68-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103195MG
Encompass_Preferred
Medchemexpress, HY-10319 5mg ROCK-IN-2 CAS:867017-68-3 ROCK-IN-2 (Azaindole 1; TC-S 7001) is an orally active and ATP-competitive ROCK inhibitor with IC50s of 0.6 and 1.1 nM for human ROCK-1 and ROCK-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8054